missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) acetate sesquihydrate, 99.9% (REO), Thermo Scientific Chemicals

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols

£71.15 - £248.24

Chemical Identifiers

CAS 537-00-8
Molecular Formula C6H9CeO6
Molecular Weight (g/mol) 317.248
MDL Number MFCD00051060
InChI Key VGBWDOLBWVJTRZ-UHFFFAOYSA-K
Synonym cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate
PubChem CID 10826
IUPAC Name cerium(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307118
View Documents
Thermo Scientific Alfa Aesar
015294.22
100 g
£71.15
100g
Please to purchase this item. Need a web account? Register with us today!
11317118
View Documents
Thermo Scientific Alfa Aesar
015294.36
500 g
£248.24
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.Cerium(III) acetate sesquihydrate, acts as a source of cerium involved in the preparation of cerium oxide. It finds application in metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. It is utilized in steel production to remove the free sulfur and oxygen. Further, it is an efficient glass polishing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

537-00-8
317.248
VGBWDOLBWVJTRZ-UHFFFAOYSA-K
10826
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]
C6H9CeO6
MFCD00051060
cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate
cerium(3+);triacetate

Specifications

308°C (decomposition)
White
C6H9CeO6
cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate
VGBWDOLBWVJTRZ-UHFFFAOYSA-K
cerium(3+);triacetate
10826
99.9%
Cerium(III) acetate sesquihydrate
537-00-8
Powder
MFCD00051060
Soluble in water.
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]
317.248
344.28 (317.26 Anhydrous)
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

missing translation for 'einecsNumber' : 208-654-0

missing translation for 'rtecsNumber' : FK4900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only