missing translation for 'onlineSavingsMsg'
Learn More
Cerium(III) acetate sesquihydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11351079

Cerium(III) acetate sesquihydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11351079
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11351079

Brand: Thermo Scientific Alfa Aesar 011324.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Specialty chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-00-8
Molecular Formula C6H9CeO6
Molecular Weight (g/mol) 317.248
MDL Number MFCD00051060
InChI Key VGBWDOLBWVJTRZ-UHFFFAOYSA-K
Synonym cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrateShow MoreShow Less
PubChem CID 10826
IUPAC Name cerium(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3]

Specifications

Melting Point 308°C (decomposition)
Physical Form Crystalline Aggregates
Quantity 250 g
Solubility Information Soluble in water (265g/L) at 15°C.
Formula Weight 344.28 (317.26 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Assay (REO)
Chemical Name or Material Cerium(III) acetate sesquihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06J037 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number011324.30,011324.09,011324.18
Lot NumberW09F020 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011324.30,011324.09,011324.18
Lot NumberD04T031 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011324.30,011324.09,011324.18
Lot NumberF1078 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011324.30,011324.09,011324.18
Lot NumberA03Y024 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011324.30,011324.09,011324.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(III) acetate sesquihydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.