Learn More
Cerium(III) acetate sesquihydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Specialty chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 308°C (decomposition) |
Physical Form | Crystalline Aggregates |
Quantity | 10 g |
Solubility Information | Soluble in water (265g/L) at 15°C. |
Formula Weight | 344.28 (317.26 Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Assay | (REO) |
Chemical Name or Material | Cerium(III) acetate sesquihydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W06J037 | Certificate of Analysis | 25/09/2023 | 011324.30,011324.09,011324.18 |
W09F020 | Certificate of Analysis | 12/11/2021 | 011324.30,011324.09,011324.18 |
D04T031 | Certificate of Analysis | 12/11/2021 | 011324.30,011324.09,011324.18 |
F1078 | Certificate of Analysis | 12/11/2021 | 011324.30,011324.09,011324.18 |
A03Y024 | Certificate of Analysis | 12/11/2021 | 011324.30,011324.09,011324.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.