Learn More
Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals
Description
A useful oxidant for organic synthesis. This chemical is also useful for the synthesis of quinoxaline derivatives which are useful as DNA cleaving agents, dyes, and organic semiconductors. Used to release organic ligands from metal carbonyls. Whereby the metal is oxidized, Carbon monoxide is produced, and the organic ligand is released.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 500 g |
Solubility Information | Soluble in water. |
Formula Weight | 558.28 (486.22 Anhydrous) |
Grade | Reagent |
Appearance | White crystalline |
Chemical Name or Material | Cerium(III) ammonium nitrate tetrahydrate, For reagent grade |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T25J029 | Certificate of Analysis | 14/08/2023 | 038367.36,038367.22 |
N11I018 | Certificate of Analysis | 12/04/2022 | 038367.36,038367.22 |
Y07E055 | Certificate of Analysis | 08/12/2021 | 038367.36,038367.22 |
W15H006 | Certificate of Analysis | 08/12/2021 | 038367.36,038367.22 |
R24E006 | Certificate of Analysis | 07/12/2021 | 038367.36,038367.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.