missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) bromide hydrate, 99%

Product Code. 11480750
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11480750

Brand: Thermo Scientific Alfa Aesar 044447.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14457-87-5 | Br3Ce | 379.83 g/mol

Cerium(III) bromide hydrate is used in the security, medical imaging, and geophysics detectors, γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14457-87-5
Molecular Formula Br3Ce
Molecular Weight (g/mol) 379.83
MDL Number MFCD00016004
InChI Key MOOUSOJAOQPDEH-UHFFFAOYSA-K
IUPAC Name cerium(3+) tribromide
SMILES [Br-].[Br-].[Br-].[Ce+3]

Specifications

Physical Form Crystalline
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 379.83 (Anhydrous)
Percent Purity 99%
Odor Odorless
Chemical Name or Material Cerium (III) bromide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.