missing translation for 'onlineSavingsMsg'
Learn More
Cerium(III) carbonate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10033872

Cerium(III) carbonate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10033872
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10033872

Brand: Thermo Scientific Acros 379400500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54451-25-1
Molecular Formula C3Ce2O9
Molecular Weight (g/mol) 460.26
MDL Number MFCD00149635
InChI Key GHLITDDQOMIBFS-UHFFFAOYSA-H
Synonym cerium iii carbonate hydrate, acmc-20alhm, cerous carbonate hydrate, ksc267s0r, ce2 co3 3.xh2o, cerium iii carbonate hydrate, reacton, cerium 3+ carbonate-water 2/3/1, dicerium 3+ hydrate tricarbonate, cerium iii carbonate hydrate, reacton reoShow MoreShow Less
PubChem CID 16212049
IUPAC Name cerium(3+);tricarbonate;hydrate
SMILES [Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99.9%
Linear Formula Ce(CO3)3·xH2O
Solubility Information Solubility in water: insoluble
Formula Weight 460.27
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Cerium(III) carbonate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0444584 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number379402500,379400500,379400100
Lot NumberA0432718 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number379402500,379400500,379400100
Lot NumberA0427247 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number379402500,379400500,379400100
Lot NumberA0425083 Certificate TypeCertificate of Analysis Date10/10/2023 Catalog Number379402500,379400500,379400100
Lot NumberA0453238 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number379402500,379400500,379400100

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(III) carbonate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.