missing translation for 'onlineSavingsMsg'
Learn More
Cerium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11327118

Cerium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11327118
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11327118

Brand: Thermo Scientific Alfa Aesar 015295.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Raw material of various high purity cerium complexes and high grad polishing powder. Cerium Carbonate is used to decolorize glass by keeping iron in its ferrous state.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54451-25-1
Molecular Formula C3Ce2O9
Molecular Weight (g/mol) 460.26
MDL Number MFCD00149635
InChI Key GHLITDDQOMIBFS-UHFFFAOYSA-H
Synonym cerium iii carbonate hydrate, acmc-20alhm, cerous carbonate hydrate, ksc267s0r, ce2 co3 3.xh2o, cerium iii carbonate hydrate, reacton, cerium 3+ carbonate-water 2/3/1, dicerium 3+ hydrate tricarbonate, cerium iii carbonate hydrate, reacton reoShow MoreShow Less
PubChem CID 16212049
SMILES [Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 848°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99.9%
Solubility Information Insoluble in water. Soluble indilute mineral acids.
IUPAC Name dicerium(3+) tricarbonate
Formula Weight 460.27 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cerium(III) carbonate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT18I036 Certificate TypeCertificate of Analysis Date31/10/2022 Catalog Number015295.36,015295.22
Lot NumberM17I037 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number015295.36,015295.22
Lot NumberU13H005 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015295.36,015295.22
Lot NumberR18H045 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015295.36,015295.22
Lot NumberQ21H044 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015295.36,015295.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.