missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) carbonate hydrate, REacton™, 99.9% (REO)

Product Code. p-4824657
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11327118

Brand: Thermo Scientific Alfa Aesar 015295.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54451-25-1 | C3Ce2O9 | 460.26 g/mol

Raw material of various high purity cerium complexes and high grad polishing powder. Cerium Carbonate is used to decolorize glass by keeping iron in its ferrous state.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54451-25-1
Molecular Formula C3Ce2O9
Molecular Weight (g/mol) 460.26
MDL Number MFCD00149635
InChI Key GHLITDDQOMIBFS-UHFFFAOYSA-H
Synonym cerium iii carbonate hydrate, acmc-20alhm, cerous carbonate hydrate, ksc267s0r, ce2 co3 3.xh2o, cerium iii carbonate hydrate, reacton, cerium 3+ carbonate-water 2/3/1, dicerium 3+ hydrate tricarbonate, cerium iii carbonate hydrate, reacton reo
PubChem CID 16212049
SMILES [Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 848°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99.9%
Solubility Information Insoluble in water. Soluble indilute mineral acids.
IUPAC Name dicerium(3+) tricarbonate
Formula Weight 460.27 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cerium(III) carbonate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.