missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride, anhydrous, 99.5% (REO), Thermo Scientific Chemicals

For Incandescent Gas Mantles, Spectrography, Polymerization of Olefins

£80.80 - £1002.99

Chemical Identifiers

CAS 7790-86-5
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.466
MDL Number MFCD00010929
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
PubChem CID 24636
ChEBI CHEBI:35458
IUPAC Name cerium(3+);trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317997
View Documents
Thermo Scientific Alfa Aesar
021112.09
10 g
£80.80
10g
Please to purchase this item. Need a web account? Register with us today!
11307997
View Documents Promotion Details
Thermo Scientific Alfa Aesar
021112.18
50 g
£256.28
50g
Please to purchase this item. Need a web account? Register with us today!
11327997
View Documents
Thermo Scientific Alfa Aesar
021112.30
250 g
£1002.99
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerium(III) chloride is used in incandescent gas mantles, spectrography and polymerization of olefins. It acts as a Lewis acid and used in Friedel-Crafts alkylation and acylation reactions in organic synthesis. It is used as a precursor for the preparation of other cerium salts, such as cerium(III) trifluoromethanesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7790-86-5
246.466
VYLVYHXQOHJDJL-UHFFFAOYSA-K
24636
cerium(3+);trichloride
CeCl3
MFCD00010929
cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
CHEBI:35458
[Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

1727°C
7790-86-5
Powder
CeCl3
cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
VYLVYHXQOHJDJL-UHFFFAOYSA-K
cerium(3+);trichloride
24636
246.48
Hygroscopic
3.92 g/cm3 at 20°C
848°C
White
99.5%
MFCD00010929
Soluble in water,alcohol,acetone,acids. Soluble in water,ethanol and acetone.
[Cl-].[Cl-].[Cl-].[Ce+3]
246.466
CHEBI:35458
Odorless
White powder
Cerium(III) chloride
Videos
Safety and Handling

Safety and Handling

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 232-227-8

missing translation for 'rtecsNumber' : FK5075000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Cerium(III) chloride anhydrous

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Cerium(III) chlorideanhydrous

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.