missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 848.0°C |
Color | Yellow |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 100 g |
Assay Percent Range | 98.5% min. (Argentometry) |
Linear Formula | CeCl3·7H2O |
Fieser | 01,121; 09,99; 15,72; 16,4; 17,314 |
Merck Index | 10, 1956 |
Solubility Information | Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol |
Formula Weight | 372.58 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0459772 | Certificate of Analysis | 07/02/2024 | 211821000,211820250,211825000 |
A0453832 | Certificate of Analysis | 25/08/2023 | 211821000,211820250,211825000 |
A0395662 | Certificate of Analysis | 15/12/2022 | 211821000,211820250,211825000 |
A0403544 | Certificate of Analysis | 15/12/2022 | 211821000,211820250,211825000 |
A0412289 | Certificate of Analysis | 13/12/2022 | 211821000,211820250,211825000 |
Safety and Handling


Product Identifier
- Cerium(III) chloride heptahydrate
- Danger
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Cerium chloride heptahydrate, Cerium chloride (CeCl3)
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cerium(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction