missing translation for 'onlineSavingsMsg'
Learn More
Cerium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11333867

Cerium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11333867
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11333867

Brand: Thermo Scientific Alfa Aesar 035688.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cerium(III) chloride is used in incandescent gas mantles, spectrography and polymerization of olefins. It acts as a Lewis acid and used in Friedel-Crafts alkylation and acylation reactions in organic synthesis. It is used as a precursor for the preparation of other cerium salts, such as cerium(III) trifluoromethanesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-86-5
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.466
MDL Number MFCD00010929
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
PubChem CID 24636
ChEBI CHEBI:35458
IUPAC Name cerium(3+);trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

Boiling Point 1727°C
Melting Point 848°C
Color Gray to White
Physical Form Crystalline
Quantity 25 g
Merck Index 14,1997
Solubility Information Soluble in water,ethanol,acids and acetone.
Formula Weight 246.48
Percent Purity 99.9%
Odor Odorless
Packaging Ampouled under Argon
Sensitivity Hygroscopic
Density 3.92
Chemical Name or Material Cerium(III) chloride, ultra dry, (REO)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX09K032 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number035688.06,035688.22,035688.14
Lot NumberR08K009 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number035688.06,035688.22,035688.14
Lot NumberR08K012 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number035688.06,035688.22,035688.14
Lot NumberR08J039 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number035688.06,035688.22,035688.14
Lot NumberM04K012 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog Number035688.06,035688.22,035688.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(III) chloride, ultra dry, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.