Learn More
Cerium(III) Nitrate, Extra Pure, SLR, Low In Other Rare Earths, Fisher Chemical
Brand: Fisher Chemical C/3780/48
Description

Specifications
10294-41-4 | |
Solid | |
CeN3O9·6H2O | |
434.22g/mol |
White | |
100 g | |
149631 | |
Plastic powder jar |
Safety and Handling
- Cerium(III) nitrate hexahydrate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.