missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) nitrate hexahydrate, 99.5%, Thermo Scientific Chemicals

Product Code. 11964371
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-41-4
Molecular Formula CeH12N3O15
Molecular Weight (g/mol) 434.22
MDL Number MFCD00149631
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
Synonym cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f
PubChem CID 16211466
SMILES O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Colorless to White
Physical Form Crystalline Powder or Agglomerating Crystals
Quantity 2.5 kg
Assay Percent Range 99.4% min. (CeO2/TREO basis)
Linear Formula Ce(NO3)3·6H2O
Fieser 16,67
Merck Index 10,1959
Solubility Information Solubility in water: 1754g/L (25°C). Other solubilities: soluble in alcohol and acetone
IUPAC Name cerium(3+) hexahydrate trinitrate
Formula Weight 434.22
Percent Purity 99.5%
Packaging Plastic bottle
Chemical Name or Material Cerium(III) nitrate hexahydrate, 99.5%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cerium(III) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'