missing translation for 'onlineSavingsMsg'
Learn More
Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals Product Code.: 11381089

Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals

Product Code. 11381089
500 g
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 11381089

Brand: Thermo Scientific Alfa Aesar 011329.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Separation of Cerium From Other Rare Earths, Catalyst For Hydrolysis of Phosporic Acid Esters

Cerium(III) nitrate hexahydrate is used in the separation of cerium from other rare earths. It plays an important role as a catalyst for hydrolysis of phosphoric acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-41-4
Molecular Formula CeH12N3O15
Molecular Weight (g/mol) 434.22
MDL Number MFCD00149631
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
Synonym cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5fShow MoreShow Less
PubChem CID 16211466
IUPAC Name cerium(3+) hexahydrate trinitrate
SMILES O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

pH 3
Physical Form Crystalline Aggregates
Quantity 500 g
Linear Formula Ce(NO3)3·6H2O
UN Number UN1477
Merck Index 14,2000
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 434.23 (326.13 Anhydrous)
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cerium(III) nitrate hexahydrate, REacton™, (REO)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT01K024 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog Number011329.36,011329.22,011329.A1,011329.09
Lot NumberT24J039 Certificate TypeCertificate of Analysis Date10/05/2024 Catalog Number011329.36,011329.22,011329.A1,011329.09
Lot NumberR10J002 Certificate TypeCertificate of Analysis Date16/05/2023 Catalog Number011329.36
Lot NumberR10J003 Certificate TypeCertificate of Analysis Date16/05/2023 Catalog Number011329.36
Lot NumberQ03J005 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number011329.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.