missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
250 g
Description
Cerium(III) nitrate hexahydrate is used in the separation of cerium from other rare earths. It plays an important role as a catalyst for hydrolysis of phosphoric acid esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Crystalline Aggregates |
Quantity | 5 g |
Assay Percent Range | 99.99% (REO) |
Solubility Information | Soluble in water,alcohol and acetone. |
Formula Weight | 434.23 (326.13 Anhydrous) |
Sensitivity | Hygroscopic |
Chemical Name or Material | Cerium(III) nitrate, hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'