Learn More
Cerium(III) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
£54.17 - £103.52
Chemical Identifiers
CAS | 14017-47-1 |
---|---|
Molecular Formula | CeCl3O12 |
Molecular Weight (g/mol) | 438.454 |
MDL Number | MFCD00150205 |
InChI Key | LJBTWTBUIINKRU-UHFFFAOYSA-K |
Synonym | cerium 3+ perchlorate, cerium perchlorate, cerous perchlorate, cerium 3+ triperchlorate, perchloric acid, cerium 3+ salt, perchloric acid, cerium 3+ salt 3:1, acmc-1bs3z, cerium iii perchlorate solution, perchloric acid cerium iii salt, perchloric acid,cerium 3+ salt 3:1Show More |
PubChem CID | 166978 |
IUPAC Name | cerium(3+);triperchlorate |
SMILES | [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ce+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311768
|
Thermo Scientific Alfa Aesar
011654.18 |
50 g |
£54.17
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301768
|
Thermo Scientific Alfa Aesar
011654.30 |
250 g |
£103.52
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14017-47-1 | |
438.454 | |
LJBTWTBUIINKRU-UHFFFAOYSA-K | |
166978 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ce+3] |
CeCl3O12 | |
MFCD00150205 | |
cerium 3+ perchlorate, cerium perchlorate, cerous perchlorate, cerium 3+ triperchlorate, perchloric acid, cerium 3+ salt, perchloric acid, cerium 3+ salt 3:1, acmc-1bs3z, cerium iii perchlorate solution, perchloric acid cerium iii salt, perchloric acid,cerium 3+ salt 3:1Show More | |
cerium(3+);triperchlorate |
Specifications
14017-47-1 | |
CeCl3O12 | |
UN1481 | |
LJBTWTBUIINKRU-UHFFFAOYSA-K | |
cerium(3+);triperchlorate | |
166978 | |
Reagent | |
Moisture sensitive |
Crystalline | |
MFCD00150205 | |
cerium 3+ perchlorate, cerium perchlorate, cerous perchlorate, cerium 3+ triperchlorate, perchloric acid, cerium 3+ salt, perchloric acid, cerium 3+ salt 3:1, acmc-1bs3z, cerium iii perchlorate solution, perchloric acid cerium iii salt, perchloric acid,cerium 3+ salt 3:1 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ce+3] | |
438.454 | |
546.56 (438.47 Anhydrous) | |
Odorless | |
Cerium(III) perchlorate hexahydrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 237-836-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Cerium(III) perchlorate hexahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Cerium(III) perchlorate hexahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.