missing translation for 'onlineSavingsMsg'
Learn More
Cerium(III) sulfate octahydrate, 99.99%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10547563

Cerium(III) sulfate octahydrate, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10547563
50 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10547563

Brand: Thermo Scientific Acros 192790500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-59-6
Molecular Formula Ce2O12S3·8H2O
Molecular Weight (g/mol) 712.55
MDL Number MFCD00149641

Specifications

Color White
Physical Form Crystals and/or Chunks
Quantity 50 g
Assay Percent Range 38.0 to 40.5% (Ce)
Linear Formula Ce2(SO4)3·8H2O
Fieser 09,99
Merck Index 10, 1961
Solubility Information Solubility in water: slightly soluble
Formula Weight 712.55
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Cerium(III) sulfate octahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465391 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number192792500,192790500
Lot NumberA0463776 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number192792500,192790500
Lot NumberA0406123 Certificate TypeCertificate of Analysis Date30/08/2023 Catalog Number192792500,192790500
Lot NumberA0405179 Certificate TypeCertificate of Analysis Date04/08/2020 Catalog Number192792500,192790500
compliance-icons
Product Identifier
  • Cerium(III) sulfate octahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(III) sulfate octahydrate, 99.99%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.