missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) sulfide, 99.9% (metals basis)

Product Code. 11394817
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11394817

Brand: Thermo Scientific Alfa Aesar 041856.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12014-93-6 | Ce2S3 | 376.41 g/mol

Used as pharmaceutical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12014-93-6
Molecular Formula Ce2S3
Molecular Weight (g/mol) 376.41
MDL Number MFCD00016007
InChI Key MMXSKTNPRXHINM-UHFFFAOYSA-N
Synonym cerium sulfide, dicerium trisulphide, ceric sulfide, cerium 3+ trisulfide, dicerium 3+ trisulfide, dicerium 3+ trisulfanediide
PubChem CID 159394
SMILES [S--].[S--].[S--].[Ce+3].[Ce+3]

Specifications

Melting Point 2100°C
Color Red to Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Insoluble in water.
IUPAC Name cerium(3+);trisulfide
Formula Weight 376.43
Odor Rotten eggs
Appearance Red-brown powder
Density 5.02 g/cm3 at 20°C
Chemical Name or Material Cerium(III) sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.