missing translation for 'onlineSavingsMsg'
Learn More
Cerium(IV) ammonium nitrate, ACS, 98.5% min, Thermo Scientific Chemicals Product Code.: 11387558

Cerium(IV) ammonium nitrate, ACS, 98.5% min, Thermo Scientific Chemicals

Product Code. 11387558
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11387558

Brand: Thermo Scientific Alfa Aesar 033254.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins

Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins, and as a scavenger in the manufacture of azides. Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16774-21-3
Molecular Formula (NH4)2Ce(NO3)6
MDL Number MFCD00151121

Specifications

Physical Form Granules
Quantity 100 g
Assay Percent Range 98.5%
Solubility Information Soluble in water and alcohol.
Formula Weight 548.23
Grade ACS
Chemical Name or Material Cerium(IV) ammonium nitrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62500036 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number033254.36,033254.22,033254.A3
Lot NumberT11K008 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number033254.36,033254.22,033254.A3
Lot NumberN09K010 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog Number033254.36,033254.22,033254.A3
Lot NumberN06E007 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number033254.36,033254.22,033254.A3
Lot NumberH28K04 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number033254.36,033254.22,033254.A3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(IV) ammonium nitrate, ACS, 98.5% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.