missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, 96% (REO)

Product Code. p-7051160
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11368548

Brand: Thermo Scientific Alfa Aesar 040581.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1306-38-3 | CeO2 | 172.114 g/mol

Cerium(IV) oxide is used in polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.114
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O

Specifications

Melting Point 2600°C
Color Yellow
Physical Form Powder
Quantity 250 g
Merck Index 14,1989
Solubility Information Insoluble in water; Soluble in H2SO4 and HNO3
Formula Weight 172.12
Percent Purity 96%
Odor Odorless
Density 7.132 g/mL
Assay (REO)
Chemical Name or Material Cerium(IV) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.