missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

£34.47 - £416.07

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.12
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10470281
View Documents
Thermo Scientific Acros
199121000
100 g Plastic bottle
£34.47
100g
Please to purchase this item. Need a web account? Register with us today!
10571451
View Documents
Thermo Scientific Acros
199125000
500 g Plastic bottle
£108.54
500g
Please to purchase this item. Need a web account? Register with us today!
13438448
View Documents
Thermo Scientific Acros
199120025
2.5 kg Plastic bottle
£416.07
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

1306-38-3
172.12
CETPSERCERDGAM-UHFFFAOYSA-N
73963
dioxocerium
CeO2
MFCD00010933
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CHEBI:79089
O=[Ce]=O

Specifications

2600.0°C
White to Yellow
99.9%
CeO2
06,99
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CETPSERCERDGAM-UHFFFAOYSA-N
dioxocerium
73963
172.12
Trace Metal Basis
7.1300g/mL
1306-38-3
Powder
CeO2
MFCD00010933
15,1988
Solubility in water: insoluble. Other solubilities: soluble in acids
O=[Ce]=O
172.12
CHEBI:79089
99.9%
7.13
Cerium(IV) oxide, 99.9%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-150-4

missing translation for 'rtecsNumber' : FK6310000

missing translation for 'tsca' : TSCA

Product Identifier
  • Cerium(IV) oxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only