missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(IV) oxide, doped with 18% zirconium, 13% in H2O, nanoparticle dispersion, high pH, <5.0nm APS Number Weighted, Thermo Scientific™
£132.00 - £525.00
Chemical Identifiers
CAS | 1306-38-3 |
---|---|
Molecular Formula | CeO2 |
Molecular Weight (g/mol) | 172.114 |
MDL Number | MFCD00010933 |
InChI Key | CETPSERCERDGAM-UHFFFAOYSA-N |
Synonym | ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline |
PubChem CID | 73963 |
ChEBI | CHEBI:79089 |
IUPAC Name | dioxocerium |
SMILES | O=[Ce]=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1306-38-3 | |
172.114 | |
CETPSERCERDGAM-UHFFFAOYSA-N | |
73963 | |
dioxocerium |
CeO2 | |
MFCD00010933 | |
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opalineShow More | |
CHEBI:79089 | |
O=[Ce]=O |
Specifications
1306-38-3 | |
High | |
CeO2 | |
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline | |
O=[Ce]=O | |
172.114 | |
CHEBI:79089 | |
Cerium(IV) oxide, Doped with 18% zirconium, 13% in H2O, nanoparticle dispersion |
Red to Brown | |
Dispersion | |
MFCD00010933 | |
CETPSERCERDGAM-UHFFFAOYSA-N | |
dioxocerium | |
73963 | |
Odorless |
Safety and Handling
missing translation for 'tsca' : TSCA
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cerium(IV) oxide, doped with 18% zirconium, 13% in H2O, nanoparticle dispersion, high pH, <5.0nm APS Number Weighted, Thermo Scientific™