missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, NanoArc™ CE-0440, 25% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals

Catalog Number 15447305
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
1306-38-3
CeO2
172.114
MFCD00010933
CETPSERCERDGAM-UHFFFAOYSA-N
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
73963
CHEBI:79089
dioxocerium
O=[Ce]=O
This item is not returnable. View return policy
1306-38-3
CeO2
172.114
MFCD00010933
CETPSERCERDGAM-UHFFFAOYSA-N
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
73963
CHEBI:79089
dioxocerium
O=[Ce]=O

Suitable for use in ultrafine acid side polishing slurries

Cerium(IV) is suitable for use in ultrafine acid side polishing slurries.Cerium(IV) oxide is widely applied as a solid electrolyte in fuel cells. It is also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass. It is utilized to increase specific surface area and inhibit sintering as well as pore size growth at a high temperature by addition with CuO/TiO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 100 g
Solubility Information Immiscible with water.
Formula Weight 172.12
Concentration or Composition (by Analyte or Components) 25% in H2O
Odor Odorless
Chemical Name or Material Cerium (IV) oxide, NanoArc™ CE-6440, Colloidal dispersion

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'