Learn More
Cerium(IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals
£57.59 - £529.64
Chemical Identifiers
CAS | 1306-38-3 |
---|---|
Molecular Formula | CeO2 |
Molecular Weight (g/mol) | 172.114 |
MDL Number | MFCD00010933 |
InChI Key | CETPSERCERDGAM-UHFFFAOYSA-N |
Synonym | ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline |
PubChem CID | 73963 |
ChEBI | CHEBI:79089 |
IUPAC Name | dioxocerium |
SMILES | O=[Ce]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11321089
|
Thermo Scientific Alfa Aesar
011327.14 |
25 g |
£57.59
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301089
|
Thermo Scientific Alfa Aesar
011327.22 |
100 g |
£171.41
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311089
|
Thermo Scientific Alfa Aesar
011327.36 |
500 g |
£529.64
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cerium(IV) oxide is used in ceramics and to polish glass and stones. It is applied as a solid electrolyte in fuel cells. It can also be employed as a co-catalyst in a number of reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1306-38-3 | |
172.114 | |
CETPSERCERDGAM-UHFFFAOYSA-N | |
73963 | |
dioxocerium |
CeO2 | |
MFCD00010933 | |
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opalineShow More | |
CHEBI:79089 | |
O=[Ce]=O |
Specifications
2600°C | |
Yellow | |
99.99% | |
MFCD00010933 | |
Insoluble in water. | |
O=[Ce]=O | |
172.114 | |
CHEBI:79089 | |
Odorless | |
Cerium(IV) oxide |
1306-38-3 | |
Powder | |
CeO2 | |
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline | |
CETPSERCERDGAM-UHFFFAOYSA-N | |
dioxocerium | |
73963 | |
172.12 | |
7.132 g/cm3 at 20°C |
Safety and Handling
H302 : Harmful if swallowed.
missing translation for 'einecsNumber' : 215-150-4
missing translation for 'rtecsNumber' : FK6310000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.