missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) and Perchloric acid, Etchant Solution, Ce(IV) concentration 0.22N, Thermo Scientific™

Product Code. 11309767
Click to view available options
Quantity:
1 L
4 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Etchant solution for printed circuits and for chrome or nichrome photoresists

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD01866051

Specifications

Physical Form Liquid
Quantity 1 L
UN Number UN3093
Concentration or Composition (by Analyte or Components) Ce(IV) concentration 0.22N
Chemical Name or Material Cerium(IV) and Perchloric acid, Etchant Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'