missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) and Perchloric acid, Etchant Solution, Ce(IV) concentration 0.30N, Thermo Scientific™

Product Code. p-7051848
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy

Product Code. 11329767

Brand: Thermo Scientific Alfa Aesar 042109.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Higher concentration etchant solution than 0.22N for faster etching

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD01866051

Specifications

Physical Form Liquid
Quantity 1 L
UN Number UN3093
Concentration or Composition (by Analyte or Components) Ce(IV) concentration 0.30N
Chemical Name or Material Cerium(IV) and Perchloric acid, Etchant Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.