missing translation for 'onlineSavingsMsg'
Learn More
Cerium(IV) sulfate solution 0.1, (0.1N), NIST standard solution ready to use, for volumetric analysis, Fisher Chemical™ Product Code.: 10635302

Cerium(IV) sulfate solution 0.1, (0.1N), NIST standard solution ready to use, for volumetric analysis, Fisher Chemical™

Product Code. 10635302
2.5 L, Amber glass bottle
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Amber glass bottle
Bag in box
This item is not returnable. View return policy

Product Code. 10635302

Brand: Fisher Chemical J/3000/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13590-82-4
Molecular Formula CeO8S2
Molecular Weight (g/mol) 332.23
MDL Number MFCD00148852
InChI Key VZDYWEUILIUIDF-UHFFFAOYSA-J
Synonym ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
PubChem CID 159684
IUPAC Name cerium(4+);disulfate
SMILES [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Yellow
Physical Form Liquid
Quantity 2.5 L
Packaging Amber glass bottle
Vapor Pressure 14mmHg at 20°C
Density Vapor Density: 0.7
compliance-icons
Product Identifier
  • Cerium (IV) sulfate 0.1M in ca. 10% w/v sulfuric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash A§ thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(IV) sulfate solution 0.1, (0.1N), NIST standard solution ready to use, for volumetric analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.