missing translation for 'onlineSavingsMsg'
Learn More
Cerium(Iv) Sulfate Tetrahydrate, Honeywell Fluka™ Product Code.: 15663470

Cerium(Iv) Sulfate Tetrahydrate, Honeywell Fluka™

Product Code. 15663470 Shop All Honeywell Fluka Products
25 g
Click to view available options
Quantity:
25 g
500 g
This item is not returnable. View return policy

Product Code. 15663470

Brand: Honeywell-Fluka 3160625G

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10294-42-5
Molecular Formula CeH8O12S2
Molecular Weight (g/mol) 404.288
MDL Number MFCD00148852
InChI Key MEXSQFDSPVYJOM-UHFFFAOYSA-J
Synonym cerium iv sulfate tetrahydrate, ceric sulfate tetrahydrate, unii-6sax34b8je, 6sax34b8je, acmc-20ecws, ceric sulphate tetrahydrate, ksc493s0d, aronis24302, sulfuric acid, cerium 4+ salt 2:1 , tetrahydrate, cerium iv sulfate hydrateShow MoreShow Less
PubChem CID 22723557
IUPAC Name cerium(4+);disulfate;tetrahydrate
SMILES O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ce+4]

Specifications

Quantity 25 g
Linear Formula Ce(SO4)2 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 404.3g/mol
Percent Purity ≥98%
Grade puriss. p.a.
Chemical Name or Material Cerium(Iv) Sulfate Tetrahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(Iv) Sulfate Tetrahydrate, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.