missing translation for 'onlineSavingsMsg'
Learn More
Cerium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals Product Code.: 11470650

Cerium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Product Code. 11470650
100 mm
Click to view available options
Quantity:
10 mm
50 mm
100 mm
This item is not returnable. View return policy

Product Code. 11470650

Brand: Thermo Scientific Alfa Aesar 044037.DM

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mylar under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-45-1
Molecular Formula Ce
Molecular Weight (g/mol) 140.12
MDL Number MFCD00010924
InChI Key GWXLDORMOJMVQZ-UHFFFAOYSA-N
Synonym cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
PubChem CID 23974
ChEBI CHEBI:33369
IUPAC Name cerium
SMILES [Ce]

Specifications

Color Silver
UN Number UN1333
Quantity 100 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.21g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Cerium rod, ≈6.35mm (0.25 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP24L043 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog Number044037.DM,044037.DJ,044037.HF
Lot NumberZ23K021 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number044037.DM,044037.DJ,044037.HF
Lot NumberY08K037 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number044037.DM,044037.DJ,044037.HF
Lot NumberW23K010 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number044037.DM,044037.DJ,044037.HF
Lot NumberT25K025 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number044037.DM,044037.DJ,044037.HF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.