missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£45.31 - £161.00
Chemical Identifiers
CAS | 7440-45-1, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | Ce |
Molecular Weight (g/mol) | 140.12 |
MDL Number | MFCD00010924 |
InChI Key | GWXLDORMOJMVQZ-UHFFFAOYSA-N |
PubChem CID | 23974 |
ChEBI | CHEBI:33369 |
IUPAC Name | cerium |
SMILES | [Ce] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10210354
|
SPEX Certiprep
PLCE2-2Y |
125 mL | Triple-leached LDPE bottle | N/A | |||||
10681173
|
SPEX Certiprep
PLCE2-2X |
500 mL | Triple-leached LDPE bottle |
£161.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ce | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-45-1, 7697-37-2, 7732-18-5 | |
140.12 | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
CHEBI:33369 | |
[Ce] |
Ce | |
MFCD00010924 | |
23974 | |
cerium |
Specifications
7440-45-1, 7697-37-2, 7732-18-5 | |
1.0, 5.0, 94.0 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Ce | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
cerium | |
23974 | |
Assurance | |
2% HNO3 |
1.0, 5.0, 94.0 | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00010924 | |
[Ce] | |
140.12 | |
CHEBI:33369 | |
Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLCE2-2Y, 2X 10,000 PLCE2-3Y, -3X |
|
Cerium Standard |
Safety and Handling
Causes severe skin burns and eye damage
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Product intended for laboratory use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title