Learn More
Cerium tungsten oxide, 99.9% (REO), Thermo Scientific™
£108.00 - £324.00
Chemical Identifiers
CAS | 13454-74-5 |
---|---|
Molecular Formula | Ce2O9W2 |
Molecular Weight (g/mol) | 791.91 |
MDL Number | MFCD00049725 |
InChI Key | PMMOMAJHUNGVIB-UHFFFAOYSA-N |
Synonym | cerium tungsten oxide, cerium iii tungstate, dicerium 3+ tritungstate, cerium 3+ dioxido dioxo tungsten 2/3, 2ce.3wo4, cerium tungstate ce2 wo4 3, cerous tungstate, ce2 wo4 3, dicerium 3+ ion tris dioxotungstenbis olateShow More |
PubChem CID | 16213782 |
SMILES | [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Ce+3].[Ce+3].[W].[W] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13454-74-5 | |
791.91 | |
PMMOMAJHUNGVIB-UHFFFAOYSA-N | |
16213782 |
Ce2O9W2 | |
MFCD00049725 | |
cerium tungsten oxide, cerium iii tungstate, dicerium 3+ tritungstate, cerium 3+ dioxido dioxo tungsten 2/3, 2ce.3wo4, cerium tungstate ce2 wo4 3, cerous tungstate, ce2 wo4 3, dicerium 3+ ion tris dioxotungstenbis olateShow More | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Ce+3].[Ce+3].[W].[W] |
Specifications
1089°C | |
Powder | |
MFCD00049725 | |
PMMOMAJHUNGVIB-UHFFFAOYSA-N | |
dicerium(3+) ditungsten nonaoxidandiide | |
16213782 | |
99.9% | |
(REO) |
13454-74-5 | |
Ce2O9W2 | |
cerium tungsten oxide, cerium iii tungstate, dicerium 3+ tritungstate, cerium 3+ dioxido dioxo tungsten 2/3, 2ce.3wo4, cerium tungstate ce2 wo4 3, cerous tungstate, ce2 wo4 3, dicerium 3+ ion tris dioxotungstenbis olate | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Ce+3].[Ce+3].[W].[W] | |
791.91 | |
1023.78 | |
Odorless | |
Cerium tungsten oxide |
Safety and Handling
GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
missing translation for 'einecsNumber' : 236-639-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.