missing translation for 'onlineSavingsMsg'
Learn More

SureSTART CERT 96-Well Plate

CERT 96-well plate constructed of polypropylene.

Brand:  SureSTART 60180-P228

Additional Details : Weight : 1.20000kg

Product Code. 17717173

  • £452.61 / Pack of 50
Estimated Shipment: 22-08-2024
to see stock.
Add to basket

Description

Description

  • 96-Well
  • U-Shaped Base
  • Polypropylene
Specifications

Specifications

Without Lid
Webseal
Yes
Full
96
1 mL
50 Pk.
127.76 x 85.48 x 33 mm
Autoclavable
Clear
HPLC GC Autosampler
Polypropylene
Non-sterile
Round
No
7 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers