missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Certified Buffer Solution, pH 10, Mettler Toledo™
Click to view available options
Quantity: 6 x 250mL
Specifications
Color | Colorless |
pH | 10 |
Physical Form | Liquid |
Quantity | 250 mL |
Solution Type | pH Buffer |
Odor | None |
Packaging | 6 Pk. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'