missing translation for 'onlineSavingsMsg'
Learn More

Certified Reference Material Density Standard, Jet Aviation Fuel, UKAS ISO 17034 / ISO 17025 Certified 250mL (nominal value: 0.79672 g/mL at 15°C), Paragon Scientific™

Density Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Density according to ASTM D4052. Alternative test methods are IP 365 and ISO 12185.

Brand:  Paragon Scientific CRMU-DEKR


Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15657498

  • £203.00 / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Fully certified in accordance with ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
  • Low levels of uncertainty achieves maximum accuracy of data
  • All data generated exclusively by UKAS ISO 17025 accredited laboratories
  • Fully traceable to international standards
  • Supplied in tamper evident UN compliant security pack
  • 12 month shelf life
  • Available from stock for immediate dispatch
  • Manufactured in the United Kingdom
Specifications

Specifications

UN 1863
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Certified Reference Material Density Standard, Jet Aviation Fuel

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P391-Collect spillage.
  • P403+P235-Store in a well-ventilated place. Keep cool.

Supplemental information
  • MIXTURE LIST-Contains : Kerosine sweetened

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Nominal values quoted. Certified Values will vary from batch to batch.