Learn More
Certified Reference Material Distillation Standard, Jet Aviation Fuel, UKAS ISO 17034 Certified 250mL (nominal value: 148.1 °C to 260.3 °C), Paragon Scientific™
Distillation Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Distillation according to ASTM D86. Alternative test methods are IP 123 and ISO 3405.
Brand:  Paragon Scientific CRMU-DIKR
Product Code. 16050050
  • £203.00 / 250mL
Explore more special offers
Description
Description
  • Fully certified in accordance with ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
  • Low levels of uncertainty achieves maximum accuracy of data
  • All data generated exclusively by UKAS ISO 17025 accredited laboratories
  • Fully traceable to international standards
  • Supplied in tamper evident UN compliant security pack
  • 12 month shelf life
  • Available from stock for immediate dispatch
  • Manufactured in the United Kingdom
  • Distillation Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Distillation according to ASTM D86. Alternative test methods are IP 123 and ISO 3405.
Specifications
Specifications
UN 1223
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Distillation Standard
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : kerosine
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted. Certified Values will vary from batch to batch.