Learn More
Certified Reference Material Freezing Point, UKAS ISO 17034, Jet Aviation Fuel, UKAS ISO 17034 Certified, 250mL (nominal value: -55.5 °C), Paragon Scientific™
Freezing Point Certified Reference Material for the verification of laboratory instruments used in testing petroleum and derivative products for Freezing Point to ASTM D2386.
Brand: Paragon Scientific CRMU-FRKR
Description
- Manufactured in accordance with ASTM / EN or IP test methods
- Certified to ISO 17034 under our UKAS accreditation
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
UN 1223 |
Safety and Handling
- Freezing Point Standard, ASTM D 2386 (-54.1 °C)
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P370+P378-In case of fire: Use dry powder or sand to extinguish.
Supplemental information
- MIXTURE LIST-Contains : kerosine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Nominal values quoted. Certified Values will vary from batch to batch.