missing translation for 'onlineSavingsMsg'
Learn More

Cesium, 99.8% (metals basis)

Product Code. 11344648
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
This item is not returnable. View return policy

Product Code. 11344648

Brand: Thermo Scientific Alfa Aesar 043005.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-46-2 | Cs | 132.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-46-2
Molecular Formula Cs
Molecular Weight (g/mol) 132.91
MDL Number MFCD00134037
InChI Key TVFDJXOCXUVLDH-UHFFFAOYSA-N
Synonym caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
PubChem CID 5354618
ChEBI CHEBI:30514
SMILES [Cs]

Specifications

Color Silver
Quantity 5 g
Assay Percent Range 99.8% (Metals basis)
IUPAC Name cesium
Chemical Name or Material Cesium
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cesium
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cesium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.