missing translation for 'onlineSavingsMsg'
Learn More

Cesium, 99.95+%, (trace metal basis), Thermo Scientific Chemicals

£126.63 - £617.07

Chemical Identifiers

CAS 7440-46-2
Molecular Formula Cs
Molecular Weight (g/mol) 132.91
MDL Number MFCD00134037
InChI Key TVFDJXOCXUVLDH-UHFFFAOYSA-N
Synonym caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
PubChem CID 5354618
ChEBI CHEBI:30514
SMILES [Cs]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10501281
View Documents
Thermo Scientific
219190010
1 g
£126.63
1g
Please to purchase this item. Need a web account? Register with us today!
10266510
View Documents
Thermo Scientific Acros
219190050
5 g
£617.07
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

7440-46-2
132.91
TVFDJXOCXUVLDH-UHFFFAOYSA-N
5354618
[Cs]
Cs
MFCD00134037
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
CHEBI:30514

Specifications

Golden
28.5°C
1.8730g/mL
Cs
15, 2007
TVFDJXOCXUVLDH-UHFFFAOYSA-N
cesium
5354618
132.91
Trace Metal Basis
7440-46-2
705.0°C
99.95+%
MFCD00134037
caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
[Cs]
132.91
CHEBI:30514
≥99.95%
Cesium, 99.95+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 231-155-4

missing translation for 'rtecsNumber' : FK9225000

missing translation for 'tsca' : TSCA

Product Identifier
  • Cesium

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only