missing translation for 'onlineSavingsMsg'
Learn More

Cesium acetate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11332018
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cesium Acetate is used in organic synthesis especially for Perkin reaction; formation of unsaturated cinnamic-type acids by the condensation of aromatic aldehydes with fatty acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.949
MDL Number MFCD00013056
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]

Specifications

Melting Point 194°C
Physical Form Crystalline
Quantity 25 g
Solubility Information Soluble in water,DMSO.
Formula Weight 191.95
Percent Purity 99.9%
Odor Weak
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'