missing translation for 'onlineSavingsMsg'
Learn More
Cesium acetate, 99%, for analysis, Thermo Scientific Chemicals Product Code.: 10433195

Cesium acetate, 99%, for analysis, Thermo Scientific Chemicals

Product Code. 10433195
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10433195

Brand: Thermo Scientific Acros 390831000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3396-11-0
Molecular Formula C2H3CsO2
Molecular Weight (g/mol) 191.96
MDL Number MFCD00013056
InChI Key ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
Synonym cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion
PubChem CID 5152919
IUPAC Name cesium;acetate
SMILES CC(=O)[O-].[Cs+]

Specifications

Melting Point 194.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 100 g
Assay Percent Range 99%
Linear Formula CH3CO2Cs
Solubility Information Solubility in water: 91.5% (21°C).
Formula Weight 191.96
Percent Purity 99%
Grade Analytical
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Cesium acetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0398029 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number390831000,390830250,390835000
Lot NumberA0403789 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number390831000,390830250,390835000
Lot NumberA0448136 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number390831000,390830250,390835000
Lot NumberA0440742 Certificate TypeCertificate of Analysis Date27/04/2022 Catalog Number390831000,390830250,390835000
Lot NumberA0430846 Certificate TypeCertificate of Analysis Date29/07/2021 Catalog Number390831000,390830250,390835000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium acetate, 99%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.