Learn More
Cesium acetate, 99% (metals basis), Thermo Scientific Chemicals
£55.68 - £191.96
Chemical Identifiers
CAS | 3396-11-0 |
---|---|
Molecular Formula | C2H3CsO2 |
Molecular Weight (g/mol) | 191.949 |
MDL Number | MFCD00013056 |
InChI Key | ZOAIGCHJWKDIPJ-UHFFFAOYSA-M |
Synonym | cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion |
PubChem CID | 5152919 |
IUPAC Name | cesium;acetate |
SMILES | CC(=O)[O-].[Cs+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334758
|
Thermo Scientific Alfa Aesar
043095.18 |
50 g |
£55.68
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344758
|
Thermo Scientific Alfa Aesar
|
250 g |
£191.96
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cesium acetate is used in organic synthesis especially in Perkin reaction to prepare unsaturated cinnamic-type acids. It is used in petroleum drilling fluids as an alternative to cesium formate. In organometallic chemistry, it is an additive for metal-ctalyzed coupling reactions. It is also used in the inversion of stereo chemistry of secondary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3396-11-0 | |
191.949 | |
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M | |
5152919 | |
CC(=O)[O-].[Cs+] |
C2H3CsO2 | |
MFCD00013056 | |
cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion | |
cesium;acetate |
Specifications
194°C | |
White | |
99% (Metal basis) | |
MFCD00013056 | |
Soluble in water and dimethyl sulfoxide. | |
CC(=O)[O-].[Cs+] | |
191.949 | |
191.95 | |
Cesium acetate |
3396-11-0 | |
Lump/Powder | |
C2H3CsO2 | |
cesium acetate, acetic acid, cesium salt, caesium acetate, acetic acid, cesium salt 1:1, acetic acid, cesiumsalt, acetic acid cesium salt, caesium 1+ ion acetate, cesiumacetate, acetic acid cesium, caesium 1+ acetate ion | |
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M | |
cesium;acetate | |
5152919 | |
Off-white crystalline powder |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H361
missing translation for 'einecsNumber' : 222-248-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only