missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium bromide, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Cesium bromide is used in optics as a beamsplitter component in wide-band spectrophotometers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 1300°C |
Melting Point | 636°C |
Color | White |
Physical Form | Crystalline |
Quantity | 250 g |
Assay Percent Range | 99% (Metals basis) |
Solubility Information | Very soluble in water. Soluble in alcohol. Insoluble in acetone |
Formula Weight | 212.81 |
Odor | Odorless |
Refractive Index | 1.6984 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'