missing translation for 'onlineSavingsMsg'
Learn More

Cesium bromide, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11386858
Click to view available options
Quantity:
50 g
250 g
7787-69-1
BrCs
212.81
MFCD00010954
LYQFWZFBNBDLEO-UHFFFAOYSA-M
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
24592
cesium;bromide
[Br-].[Cs+]
7787-69-1
BrCs
212.81
MFCD00010954
LYQFWZFBNBDLEO-UHFFFAOYSA-M
cesium bromide, caesium bromide, cesium bromide csbr, tricesium tribromide, csbr, unii-06m25edm3f, caesium 1+ ion bromide, cesiumbromide, br.cs, caesium i bromide
24592
cesium;bromide
[Br-].[Cs+]

Cesium bromide is used in optics as a beamsplitter component in wide-band spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1300°C
Melting Point 636°C
Color White
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99% (Metals basis)
Solubility Information Very soluble in water. Soluble in alcohol. Insoluble in acetone
Formula Weight 212.81
Odor Odorless
Refractive Index 1.6984
Sensitivity Hygroscopic
Density 4.44 g/cm3 at 20°C
Chemical Name or Material Cesium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'