missing translation for 'onlineSavingsMsg'
Learn More

Cesium carbonate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11331458
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
534-17-8
CCs2O3
325.82
MFCD00010957
FJDQFPXHSGXQBY-UHFFFAOYSA-L
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
10796
[Cs+].[Cs+].[O-]C([O-])=O
This item is not returnable. View return policy
534-17-8
CCs2O3
325.82
MFCD00010957
FJDQFPXHSGXQBY-UHFFFAOYSA-L
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
10796
[Cs+].[Cs+].[O-]C([O-])=O

Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY
Melting Point 610°C (decomposition)
Physical Form Powder/Granules
Quantity 5 g
Assay Percent Range 99.9% (metals basis)
Beilstein 4546405
Solubility Information Freely soluble in water. Soluble in ethanol,dimethylformamide and dimethyl sulfoxide.
IUPAC Name dicaesium(1+) carbonate
Formula Weight 325.82
Sensitivity Hygroscopic
Density 4.072
Chemical Name or Material Cesium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'