missing translation for 'onlineSavingsMsg'
Learn More

Cesium carbonate, 99% (metals basis), Thermo Scientific Chemicals

Catalyst in ethylene oxide polymerization, in coating spatter-free welding of steel in CO2, and in oxide cathodes

£34.37 - £430.14

Chemical Identifiers

CAS 534-17-8
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
Synonym cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
PubChem CID 10796
SMILES [Cs+].[Cs+].[O-]C([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11367617
View Documents
Thermo Scientific Alfa Aesar
012887.06
5 g
£34.37
5g
Please to purchase this item. Need a web account? Register with us today!
11377617
View Documents
Thermo Scientific Alfa Aesar
012887.18
50 g
£41.21
50g
Please to purchase this item. Need a web account? Register with us today!
11347617
View Documents
Thermo Scientific Alfa Aesar
012887.30
250 g
£131.66
250g
Please to purchase this item. Need a web account? Register with us today!
11357617
View Documents
Thermo Scientific Alfa Aesar
012887.A1
1 kg
£430.14
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cesium carbonate is the precursor for other cesium compounds. It acts as a base in sensitive organic reactions. Suzuki, Heck and Sonogashira synthesis are some of the important reactions involving cesium carbonate. It finds use in solar cells as it increases the power conversion efficiency of cells through the transfer of electrons. It is also used in the production of special optical glasses, petroleum catalytic additives, special ceramics, and in sulfuric acid industry. It is useful in the N-alkylation (of sulfonamides, beta-lactams, indoles, heterocycles, and several sensitive nitrogen compounds), carbamination of amines, carbonylation of alcohols, and aerobic oxidation of alcohols into carbonyl compounds without over oxidations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

534-17-8
325.82
FJDQFPXHSGXQBY-UHFFFAOYSA-L
10796
CCs2O3
MFCD00010957
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
[Cs+].[Cs+].[O-]C([O-])=O

Specifications

610°C (decomposition)
Powder/Granules
CCs2O3
4546405
Freely soluble in water. Soluble in ethanol,dimethylformamide and dimethyl sulfoxide.
[Cs+].[Cs+].[O-]C([O-])=O
325.82
325.82
4.072 g/mL
534-17-8
99% (metals basis)
MFCD00010957
cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
FJDQFPXHSGXQBY-UHFFFAOYSA-L
dicaesium(1+) carbonate
10796
Hygroscopic
Cesium carbonate
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c

H318-H361f-H373

missing translation for 'einecsNumber' : 208-591-9

missing translation for 'rtecsNumber' : FK9400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.