Learn More
Cesium carbonate, 99% (metals basis), Thermo Scientific Chemicals
Description
Cesium carbonate is the precursor for other cesium compounds. It acts as a base in sensitive organic reactions. Suzuki, Heck and Sonogashira synthesis are some of the important reactions involving cesium carbonate. It finds use in solar cells as it increases the power conversion efficiency of cells through the transfer of electrons. It is also used in the production of special optical glasses, petroleum catalytic additives, special ceramics, and in sulfuric acid industry. It is useful in the N-alkylation (of sulfonamides, beta-lactams, indoles, heterocycles, and several sensitive nitrogen compounds), carbamination of amines, carbonylation of alcohols, and aerobic oxidation of alcohols into carbonyl compounds without over oxidations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 610°C (decomposition) |
Physical Form | Powder/Granules |
Quantity | 50 g |
Assay Percent Range | 99% (metals basis) |
Beilstein | 4546405 |
Solubility Information | Freely soluble in water. Soluble in ethanol,dimethylformamide and dimethyl sulfoxide. |
IUPAC Name | dicaesium(1+) carbonate |
Formula Weight | 325.82 |
Sensitivity | Hygroscopic |
Density | 4.072 g/mL |
Show More |
Safety and Handling
- Cesium carbonate
- Danger
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H318-Causes serious eye damage.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Cesium carbonate
RUO – Research Use Only