missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, 99.9% (metals basis), Thermo Scientific Chemicals

£53.16 - £138.69

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326717
View Documents
Thermo Scientific Alfa Aesar
010018.14
25 g
£53.16
25g
Please to purchase this item. Need a web account? Register with us today!
11336717
View Documents
Thermo Scientific Alfa Aesar
010018.22
100 g
£138.69
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Analytical chemistry reagent for detecting inorganic ions. Source of cesium metal. Phase transfer catalyst in organic chemistry. Used for density gradient work to separate and purify proteins (isopycnic centrifugation) including chromosomal, plasmid and organelle (mitochondrial or plasmid) DNAs, and different RNA types. Preparation of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-17-8
168.36
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
caesium(1+) chloride
ClCs
MFCD00010955
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
CHEBI:63039
[Cl-].[Cs+]

Specifications

1303°C
7647-17-8
99.9% (metals basis)
MFCD00010955
Soluble in water and concentrated hydrochloric acid.
[Cl-].[Cs+]
168.36
CHEBI:63039
1.6418
3.99
646°C
Crystalline
ClCs
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
AIYUHDOJVYHVIT-UHFFFAOYSA-M
caesium(1+) chloride
24293
168.36
Hygroscopic
Cesium chloride
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P281-P308+P313-P501c

H361-H373

missing translation for 'einecsNumber' : 231-600-2

missing translation for 'rtecsNumber' : FK9625000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only