missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, For UV/VIS spectroscopy
Specifications
Specifications
| Boiling Point | 1290.0°C |
| Melting Point | 646.0°C |
| Color | Colorless to Yellow |
| Physical Form | Crystalline Solid |
| Quantity | 25 g |
| Assay Percent Range | 99.99% |
| Linear Formula | CsCl |
| Merck Index | 15, 2010 |
| Solubility Information | Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone |
| IUPAC Name | cesium;chloride |
| Show More |
Safety and Handling
Product Identifier
- Cesium chloride
- Warning
- Reproductive toxicity Category 2
- H361-Suspected of damaging fertility or the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction