missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, MP Biomedicals™

Product Code. 11491652 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11491652

Brand: MP Biomedicals 04813063

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cesium Chloride Ultra Pure, 99.999%, White Powder

  • Cesium chloride , an inorganic compound, is an important source of cesium ions in a variety of applications.
  • Cesium chloride is typically used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins.
  • It is used for the preparation of electrically conducting glasses, used to make solutions for the separation of RNA from DNA by density gradient centrifugation.
  • Cesium chloride (CsCl) is commonly used in the separation of DNA and RNA.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym Cesium Monochloride, Dicesium Dichloride
PubChem CID 24293
ChEBI CHEBI:63039
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C (literature)
Melting Point 646°C (literature)
Color White
pH 5.5 to 6.5 (1% aq soln)
Physical Form Powder
Quantity 500 g
Solubility Information Freely soluble in water (500mg/mL),(50% aqueous-clear, colorless solution); soluble in ethanol, alcohol
Grade 0.99999
Refractive Index 1.642(Lit.)
Density 3.99g/mL(Lit.)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.