missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, Puriss. p.a., ≥99.5%, Honeywell Fluka™

Catalog Number 15683510
Click to view available options
Quantity:
100 g
6 x 100 g
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]

Puriss. p.a.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 100 g
Linear Formula CsCl
UN Number NONH for all modes of transport
Formula Weight 168.36g/mol
Percent Purity ≥99.5%
Grade puriss. p.a.
Packaging Poly bottle
Chemical Name or Material Cesium Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'